Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501039.2 | XP_003501087.1 | 2 - 3 | 276 | 139 | |
GenBank mRNA | JI876068.1 | - | 2 - 3 | 264 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |