Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496566.2 | XP_003496614.1 | 2 - 3 | 268 | 105 | |
GenBank mRNA | JI868866.1 | - | 1 - 2 | 153 | 105 | |
JP050971.1 | - | 1 - 2 | 153 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |