Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496566.2 | XP_003496614.1 | 4 - 5 | 613 | 86 | |
GenBank mRNA | JI868866.1 | - | 3 - 4 | 498 | 86 | |
JP050971.1 | - | 3 - 4 | 498 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |