Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Panx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506484.2 | XP_003506532.1 | 4 - 5 | 1562 | 64 | |
GenBank mRNA | JI883140.1 | - | 4 - 5 | 1183 | 64 | |
JP060882.1 | - | 4 - 5 | 1183 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |