Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507236.2 | XP_003507284.1 | 3 - 4 | 381 | 106 | |
XM_007647809.1 | XP_007645999.1 | 4 - 5 | 418 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |