Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vopp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645395.1 | XP_007643585.1 | 2 - 3 | 265 | 76 | |
GenBank mRNA | JP047410.1 | - | 1 - 2 | 50 | 76 |