Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vopp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645395.1 | XP_007643585.1 | 4 - 5 | 478 | 49 | |
GenBank mRNA | JP047410.1 | - | 3 - 4 | 263 | 49 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |