Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508745.1 | XP_003508793.1 | 2 - 3 | 347 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |