Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511529.2 | XP_003511577.1 | 6 - 7 | 503 | 64 | |
GenBank mRNA | JP041294.1 | - | 3 - 4 | 280 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |