Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609617.1 | XP_007607807.1 | 1 | 1821 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |