Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650320.1 | XP_007648510.1 | 2 - 3 | 633 | 92 | |
GenBank mRNA | JI872859.1 | - | 2 - 3 | 637 | 92 | |
JP049942.1 | - | 2 - 3 | 637 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |