Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643418.1 | XP_007641608.1 | 2 - 3 | 504 | 86 | |
XM_007643419.1 | XP_007641609.1 | 3 - 4 | 633 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Nuclear Receptor Probe spans exons |