Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501542.2 | XP_003501590.2 | 4 - 5 | 1293 | 70 | |
XM_007642196.1 | XP_007640386.1 | 4 - 5 | 1320 | 70 | ||
XM_007642198.1 | XP_007640388.1 | 4 - 5 | 1290 | 70 | ||
XM_007642199.1 | XP_007640389.1 | 5 - 6 | 582 | 70 | ||
XM_007642200.1 | XP_007640390.1 | 5 - 6 | 528 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |