Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcfl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503149.2 | XP_003503197.1 | 5 - 6 | 794 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |