Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504613.2 | XP_003504661.1 | 3 - 4 | 442 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |