Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652680.1 | XP_007650870.1 | 1 - 2 | 163 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Probe spans exons |