Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501266.2 | XP_003501314.1 | 2 - 3 | 1529 | 88 | |
XM_007641935.1 | XP_007640125.1 | 2 - 3 | 1006 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Probe spans exons |