Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100752154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507016.2 | XP_003507064.2 | 4 - 5 | 731 | 114 | |
XM_007647547.1 | XP_007645737.1 | 5 - 6 | 879 | 114 | ||
XM_007647548.1 | XP_007645738.1 | 3 - 4 | 646 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |