Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100774508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509753.2 | XP_003509801.1 | 10 - 11 | 1260 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |