Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497857.1 | XP_003497905.1 | 29 - 30 | 4724 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |