Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mansc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501259.2 | XP_003501307.1 | 2 - 3 | 660 | 60 | |
XM_007641925.1 | XP_007640115.1 | 3 - 4 | 815 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |