Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511045.2 | XP_003511093.1 | 2 - 3 | 523 | 77 | |
XM_007651608.1 | XP_007649798.1 | 3 - 4 | 583 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |