Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511045.2 | XP_003511093.1 | 3 - 4 | 669 | 73 | |
XM_003511046.2 | XP_003511094.1 | 2 - 3 | 609 | 73 | ||
XM_007651608.1 | XP_007649798.1 | 4 - 5 | 729 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |