Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496995.1 | XP_003497043.1 | 3 - 4 | 458 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |