Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503623.1 | XP_003503671.1 | 6 - 7 | 798 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |