Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc90b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507014.2 | XP_003507062.1 | 2 - 3 | 516 | 66 | |
GenBank mRNA | JI875122.1 | - | 2 - 3 | 263 | 66 | |
JP039804.1 | - | 2 - 3 | 263 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |