Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507997.2 | XP_003508045.1 | 2 - 3 | 179 | 88 | |
XM_007648623.1 | XP_007646813.1 | 2 - 3 | 403 | 88 | ||
GenBank mRNA | JI895357.1 | - | 1 - 2 | 59 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |