Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651253.1 | XP_007649443.1 | 4 - 5 | 650 | 77 | |
XM_007651254.1 | XP_007649444.1 | 4 - 5 | 650 | 77 | ||
GenBank mRNA | JI880477.1 | - | 2 - 3 | 243 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |