Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496470.1 | XP_003496518.1 | 2 - 3 | 150 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |