Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497187.2 | XP_003497235.1 | 4 - 5 | 507 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |