Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515248.2 | XP_003515296.1 | 2 - 3 | 439 | 114 | |
GenBank mRNA | JI864165.1 | - | 2 - 3 | 265 | 114 | |
JP050655.1 | - | 2 - 3 | 264 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |