Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650157.1 | XP_007648347.1 | 1 - 2 | 191 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |