Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501528.1 | XP_003501576.1 | 2 - 3 | 222 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Probe spans exons |