Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506496.1 | XP_003506544.1 | 3 - 4 | 466 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Probe spans exons |