Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499310.2 | XP_003499358.2 | 1 - 2 | 65 | 79 | |
XM_007640086.1 | XP_007638276.1 | 1 - 2 | 65 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |