Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506706.2 | XP_003506754.1 | 3 - 4 | 574 | 77 | |
XM_007647251.1 | XP_007645441.1 | 3 - 4 | 574 | 77 | ||
GenBank mRNA | JI862127.1 | - | 1 - 2 | 133 | 77 | |
JP058424.1 | - | 3 - 4 | 291 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |