Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007627137.1 | XP_007625327.1 | 1 - 2 | 47 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |