Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100774007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503446.2 | XP_003503494.1 | 4 - 5 | 789 | 98 |