Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504611.2 | XP_003504659.1 | 1 - 2 | 164 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |