Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511411.2 | XP_003511459.1 | 3 - 4 | 279 | 108 | |
GenBank mRNA | JI884779.1 | - | 2 - 3 | 171 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Probe spans exons |