Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100763131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509581.2 | XP_003509629.1 | 2 - 3 | 177 | 66 | |
GenBank mRNA | JP045829.1 | - | 1 - 2 | 86 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |