Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100763131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509581.2 | XP_003509629.1 | 5 - 6 | 588 | 94 | |
GenBank mRNA | JP045829.1 | - | 4 - 5 | 497 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |