Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513642.1 | XP_003513690.1 | 5 - 6 | 2167 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |