Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649414.1 | XP_007647604.1 | 3 - 4 | 5836 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |