Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645258.1 | XP_007643448.1 | 3 - 4 | 252 | 87 | |
GenBank mRNA | JP043095.1 | - | 3 - 4 | 252 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |