Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500463.2 | XP_003500511.1 | 1 - 2 | 59 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Detect Genomic DNA Probe spans exons |