Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500463.2 | XP_003500511.1 | 4 - 5 | 719 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Detect Genomic DNA Probe spans exons |