Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504132.2 | XP_003504180.2 | 4 - 5 | 615 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |