Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss11e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510461.1 | XP_003510509.1 | 3 - 4 | 324 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protease Probe spans exons |