Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501926.2 | XP_003501974.1 | 5 - 6 | 1341 | 90 | |
XM_007642543.1 | XP_007640733.1 | 6 - 7 | 1412 | 90 | ||
GenBank mRNA | JI874796.1 | - | 4 - 5 | 710 | 90 | |
JP047276.1 | - | 4 - 5 | 710 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |